Assay Details
| Gene Symbol: Rab34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159482.1 | NP_001152954.1 | 9 - 10 | 851 | 80 | |
| NM_033475.3 | NP_258436.2 | 8 - 9 | 1196 | 80 | ||
| GenBank mRNA | AB082927.1 | - | 8 - 9 | 1109 | 80 | |
| AB232630.1 | - | 8 - 9 | 607 | 80 | ||
| AF322068.1 | - | 8 - 9 | 607 | 80 | ||
| AF327929.1 | - | 9 - 10 | 801 | 80 | ||
| AK160308.1 | - | 1165 | 80 | |||
| BC038638.1 | - | 8 - 9 | 671 | 5058535 | 80 | |
| S72304.1 | - | 6 - 7 | 454 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab34
Genomic Map