Assay Details
| Gene Symbol: Mark3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021516.4 | NP_067491.2 | 10 - 11 | 1537 | 143 | |
| NM_022801.4 | NP_073712.2 | 10 - 11 | 1537 | 143 | ||
| GenBank mRNA | AF240782.1 | - | 10 - 11 | 1082 | 143 | |
| AF240783.1 | - | 10 - 11 | 1082 | 143 | ||
| AK002654.1 | - | 1 - 2 | 99 | 143 | ||
| AK083683.1 | - | 10 - 11 | 1538 | 143 | ||
| AK132247.1 | - | 10 - 11 | 1521 | 143 | ||
| AK166157.1 | - | 10 - 11 | 1526 | 143 | ||
| AK217056.1 | - | 2 - 3 | 198 | 143 | ||
| AK220557.1 | - | 9 - 10 | 5371 | 143 | ||
| BC026445.1 | - | 3 - 4 | 442 | 4459439 | 143 | |
| BC066071.1 | - | 10 - 11 | 1511 | 143 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark3
Genomic Map