Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 11 - 12 | 1355 | 101 | |
| GenBank mRNA | AK010811.1 | - | 4 - 5 | 427 | 102 | |
| AK015195.1 | - | 11 - 12 | 1359 | 101 | ||
| BC145646.1 | - | 9 - 10 | 1136 | 40130949 | 101 | |
| DV660077.1 | - | 2 - 3 | 219 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Patl2
Genomic Map