Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 2 - 3 | 207 | 60 | |
| GenBank mRNA | AK015195.1 | - | 2 - 3 | 208 | 60 | |
| BY714747.1 | - | 2 - 3 | 208 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015195.1, NM_026251.2 transcripts.
Set Membership: |
|

Patl2
Genomic Map