Assay Details
| Gene Symbol: Patl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026251.2 | NP_080527.2 | 8 - 9 | 989 | 69 | |
| GenBank mRNA | AK010811.1 | - | 1 - 2 | 61 | 69 | |
| AK015195.1 | - | 8 - 9 | 990 | 69 | ||
| BC145646.1 | - | 6 - 7 | 770 | 40130949 | 69 | |
| DV658440.1 | - | 6 - 7 | 600 | 69 | ||
More Information
Set Membership: |
|

Patl2
Genomic Map