Assay Details
| Gene Symbol: Duox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177610.2 | NP_808278.2 | 15 - 16 | 1947 | 77 | |
| GenBank mRNA | BC172138.1 | - | 15 - 16 | 2239 | 9054808 | 77 |
More Information
Updated Mapping Information
Set Membership: |
|

Duox2
Genomic Map