Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081006.1 | NP_001074475.1 | 13 - 14 | 2704 | 61 | |
| NM_001177630.2 | NP_001171101.2 | 12 - 13 | 2802 | 61 | ||
| NM_001177631.1 | NP_001171102.1 | 11 - 12 | 2697 | 61 | ||
| NM_029895.4 | NP_084171.2 | 12 - 13 | 2910 | 61 | ||
| NM_178059.5 | NP_835160.2 | 12 - 13 | 2910 | 61 | ||
| GenBank mRNA | AB125594.2 | - | 12 - 13 | 2910 | 61 | |
| AB125595.2 | - | 12 - 13 | 2910 | 61 | ||
| AK054453.1 | - | 7 - 8 | 1853 | 61 | ||
| AK086078.1 | - | 7 - 8 | 1855 | 61 | ||
| AK147510.1 | - | 11 - 12 | 2604 | 61 | ||
| BB040164.2 | - | 1 - 2 | 199 | 61 | ||
| BC172143.1 | - | 11 - 12 | 2417 | 61 | ||
More Information
Set Membership: |
|

Etl4
Genomic Map