Assay Details
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081006.1 | NP_001074475.1 | 15 - 16 | 3342 | 87 | |
| NM_001177630.2 | NP_001171101.2 | 14 - 15 | 3440 | 87 | ||
| NM_001177631.1 | NP_001171102.1 | 13 - 14 | 3335 | 87 | ||
| NM_029895.4 | NP_084171.2 | 14 - 15 | 3515 | 87 | ||
| NM_178059.5 | NP_835160.2 | 14 - 15 | 3548 | 87 | ||
| GenBank mRNA | AB125594.2 | - | 14 - 15 | 3548 | 87 | |
| AB125595.2 | - | 14 - 15 | 3515 | 87 | ||
| AK054453.1 | - | 9 - 10 | 2458 | 87 | ||
| AK147510.1 | - | 13 - 14 | 3243 | 87 | ||
| BC026657.1 | - | 2 - 3 | 477 | 87 | ||
| BC158051.1 | - | 14 - 15 | 3162 | 9007405 | 87 | |
| BC172143.1 | - | 13 - 14 | 3053 | 87 | ||
More Information
Set Membership: |
|

Etl4
Genomic Map