Assay Details
| Gene Symbol: Hoxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277238.1 | NP_001264167.1 | 2 - 3 | 1673 | 65 | |
| NM_010456.3 | NP_034586.1 | 1 - 2 | 1846 | 65 | ||
| GenBank mRNA | AB005457.1 | - | 1 - 2 | 1846 | 65 | |
| BC055059.1 | - | 1 - 2 | 656 | 6529502 | 65 | |
| M28449.1 | - | 1 - 2 | 52 | 65 | ||
More Information
Set Membership: |
|

Hoxa9
Genomic Map