Assay Details
| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145567.1 | NP_663542.1 | 1 - 2 | 142 | 88 | |
| GenBank mRNA | AB051067.1 | - | 1 - 2 | 203 | 88 | |
| AK078175.1 | - | 1 - 2 | 93 | 88 | ||
| BC003914.1 | - | 1 - 2 | 142 | 3484897 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hibadh
Genomic Map