Assay Details
| Gene Symbol: Dbh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138942.3 | NP_620392.2 | 1 - 2 | 361 | 73 | |
| GenBank mRNA | AK132245.1 | - | 1 - 2 | 362 | 73 | |
| BC141022.1 | - | 1 - 2 | 388 | 9055634 | 73 | |
| BC171949.1 | - | 1 - 2 | 388 | 9054235 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dbh
Genomic Map