Assay Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038700.2 | NP_001033789.1 | 10 - 11 | 1301 | 129 | |
| NM_001177648.1 | NP_001171119.1 | 12 - 13 | 1499 | 129 | ||
| NM_001177649.1 | NP_001171120.1 | 10 - 11 | 1301 | 129 | ||
| NM_001177650.1 | NP_001171121.1 | 9 - 10 | 1151 | 129 | ||
| GenBank mRNA | AK122308.1 | - | 12 - 13 | 1499 | 129 | |
| AK143512.1 | - | 9 - 10 | 1152 | 129 | ||
| AK157135.1 | - | 10 - 11 | 1292 | 129 | ||
| AK170497.1 | - | 10 - 11 | 1251 | 129 | ||
| AK172100.1 | - | 10 - 11 | 1245 | 129 | ||
| U40751.1 | - | 2 - 3 | 146 | 129 | ||
More Information
Set Membership: |
|

Fnbp1
Genomic Map