Assay Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038700.2 | NP_001033789.1 | 13 - 14 | 1694 | 67 | |
| NM_001177648.1 | NP_001171119.1 | 15 - 16 | 1892 | 67 | ||
| NM_001177649.1 | NP_001171120.1 | 13 - 14 | 1697 | 67 | ||
| NM_001177650.1 | NP_001171121.1 | 12 - 13 | 1544 | 67 | ||
| GenBank mRNA | AK122308.1 | - | 15 - 16 | 1892 | 67 | |
| AK143512.1 | - | 12 - 13 | 1545 | 67 | ||
| AK157135.1 | - | 13 - 14 | 1685 | 67 | ||
| AK170497.1 | - | 13 - 14 | 1647 | 67 | ||
| AK172100.1 | - | 13 - 14 | 1638 | 67 | ||
| U40751.1 | - | 5 - 6 | 539 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fnbp1
Genomic Map