Assay Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290625.1 | NP_001277554.1 | 11 - 12 | 1591 | 57 | |
| NM_001290626.1 | NP_001277555.1 | 10 - 11 | 1589 | 57 | ||
| NM_177345.4 | NP_796319.1 | 11 - 12 | 1917 | 57 | ||
| GenBank mRNA | AK027932.1 | - | 6 - 7 | 690 | 57 | |
| AK042154.1 | - | 10 - 11 | 1579 | 57 | ||
| AK052045.1 | - | 11 - 12 | 1585 | 57 | ||
| AK132263.1 | - | 11 - 12 | 1581 | 57 | ||
| BC027377.1 | - | 6 - 7 | 719 | 57 | ||
| BC031579.1 | - | 6 - 7 | 719 | 57 | ||
| BC043296.1 | - | 11 - 12 | 1816 | 57 | ||
| BC090644.1 | - | 10 - 11 | 1491 | 30629263 | 57 | |
| BC096618.1 | - | 11 - 12 | 1890 | 6392317 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapkap1
Genomic Map