Assay Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290625.1 | NP_001277554.1 | 4 - 5 | 649 | 84 | |
| NM_001290626.1 | NP_001277555.1 | 3 - 4 | 647 | 84 | ||
| NM_177345.4 | NP_796319.1 | 4 - 5 | 975 | 84 | ||
| GenBank mRNA | AK042154.1 | - | 3 - 4 | 637 | 84 | |
| AK052045.1 | - | 4 - 5 | 643 | 84 | ||
| AK132263.1 | - | 4 - 5 | 639 | 84 | ||
| AK148428.1 | - | 4 - 5 | 964 | 84 | ||
| BC043296.1 | - | 4 - 5 | 874 | 84 | ||
| BC090644.1 | - | 3 - 4 | 549 | 30629263 | 84 | |
| BC096618.1 | - | 4 - 5 | 948 | 6392317 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001290626.1 transcripts.
Set Membership: |
|

Mapkap1
Genomic Map