Assay Details
| Gene Symbol: Mapkap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290625.1 | NP_001277554.1 | 8 - 9 | 1215 | 94 | |
| NM_001290626.1 | NP_001277555.1 | 7 - 8 | 1213 | 94 | ||
| NM_177345.4 | NP_796319.1 | 8 - 9 | 1541 | 94 | ||
| GenBank mRNA | AK027932.1 | - | 3 - 4 | 314 | 94 | |
| AK042154.1 | - | 7 - 8 | 1203 | 94 | ||
| AK052045.1 | - | 8 - 9 | 1209 | 94 | ||
| AK132263.1 | - | 8 - 9 | 1205 | 94 | ||
| AK148428.1 | - | 8 - 9 | 1531 | 94 | ||
| BC027377.1 | - | 3 - 4 | 343 | 94 | ||
| BC031579.1 | - | 3 - 4 | 343 | 94 | ||
| BC043296.1 | - | 8 - 9 | 1440 | 94 | ||
| BC090644.1 | - | 7 - 8 | 1115 | 30629263 | 94 | |
| BC096618.1 | - | 8 - 9 | 1514 | 6392317 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mapkap1
Genomic Map