Assay Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025744.2 | NP_080020.2 | 11 - 12 | 1476 | 68 | |
| GenBank mRNA | AK006384.1 | - | 11 - 12 | 804 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erich2
Genomic Map