Assay Details
| Gene Symbol: Erich2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025744.2 | NP_080020.2 | 8 - 9 | 1088 | 109 | |
| GenBank mRNA | AK006384.1 | - | 8 - 9 | 417 | 108 | |
| AK017113.1 | - | 8 - 9 | 428 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Erich2
Genomic Map