Assay Details
| Gene Symbol: Kif18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139303.1 | NP_647464.1 | 13 - 14 | 2149 | 70 | |
| GenBank mRNA | AK044795.1 | - | 13 - 14 | 2044 | 70 | |
| AK049197.1 | - | 13 - 14 | 2099 | 70 | ||
| AK164649.1 | - | 13 - 14 | 2044 | 70 | ||
| BC016095.1 | - | 13 - 14 | 2149 | 4506052 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif18a
Genomic Map