Assay Details
| Gene Symbol: Kif18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139303.1 | NP_647464.1 | 15 - 16 | 2674 | 78 | |
| GenBank mRNA | AK044795.1 | - | 15 - 16 | 2569 | 78 | |
| AK049197.1 | - | 15 - 16 | 2624 | 78 | ||
| AK164649.1 | - | 15 - 16 | 2569 | 78 | ||
| BC016095.1 | - | 15 - 16 | 2674 | 4506052 | 78 | |
More Information
Set Membership: |
|

Kif18a
Genomic Map