Assay Details
| Gene Symbol: Kif18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139303.1 | NP_647464.1 | 2 - 3 | 527 | 78 | |
| GenBank mRNA | AK036722.1 | - | 2 - 3 | 414 | 78 | |
| AK041469.1 | - | 2 - 3 | 426 | 78 | ||
| AK044795.1 | - | 2 - 3 | 422 | 78 | ||
| AK049197.1 | - | 2 - 3 | 476 | 78 | ||
| AK082913.1 | - | 2 - 3 | 440 | 78 | ||
| AK135735.1 | - | 2 - 3 | 447 | 78 | ||
| AK164114.1 | - | 2 - 3 | 397 | 78 | ||
| AK164649.1 | - | 2 - 3 | 422 | 78 | ||
| BC004740.1 | - | 1 - 2 | 382 | 78 | ||
| BC016095.1 | - | 2 - 3 | 527 | 4506052 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif18a
Genomic Map