Assay Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290503.1 | NP_001277432.1 | 10 - 11 | 2030 | 74 | |
| NM_178694.4 | NP_848809.2 | 10 - 11 | 2069 | 74 | ||
| GenBank mRNA | AK155401.1 | - | 10 - 11 | 1977 | 74 | |
| BC048924.1 | - | 10 - 11 | 1912 | 6312691 | 74 | |
| BC057021.1 | - | 10 - 11 | 1974 | 6825937 | 74 | |
More Information
Set Membership: |
|

Zer1
Genomic Map