Assay Details
| Gene Symbol: Zer1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290503.1 | NP_001277432.1 | 4 - 5 | 1156 | 79 | |
| GenBank mRNA | AK029290.1 | - | 4 - 5 | 1130 | 79 | |
| AK032153.1 | - | 4 - 5 | 1126 | 79 | ||
| AK032318.1 | - | 3 - 4 | 802 | 79 | ||
| AK035712.1 | - | 1 - 2 | 108 | 79 | ||
| AK048853.1 | - | 4 - 5 | 1051 | 79 | ||
| AK081355.1 | - | 4 - 5 | 1033 | 79 | ||
| AK087046.1 | - | 4 - 5 | 1029 | 79 | ||
| AK155401.1 | - | 4 - 5 | 1103 | 79 | ||
| BC057021.1 | - | 4 - 5 | 1100 | 6825937 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zer1
Genomic Map