Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 10 - 11 | 1134 | 77 | |
| GenBank mRNA | AK031511.1 | - | 10 - 11 | 1147 | 77 | |
| AK042391.1 | - | 9 - 10 | 1080 | 77 | ||
| AK088404.1 | - | 9 - 10 | 1042 | 77 | ||
| AK163165.1 | - | 9 - 10 | 942 | 77 | ||
| BC016206.1 | - | 10 - 11 | 1236 | 4912335 | 77 | |
| BC025124.1 | - | 9 - 10 | 1267 | 4486058 | 77 | |
| BC052047.1 | - | 10 - 11 | 1134 | 5687475 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map