Assay Details
| Gene Symbol: Cercam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207298.2 | NP_997181.1 | 4 - 5 | 598 | 97 | |
| GenBank mRNA | AK155384.1 | - | 4 - 5 | 594 | 97 | |
| BC052176.1 | - | 4 - 5 | 559 | 97 | ||
| BC138847.1 | - | 4 - 5 | 577 | 8861869 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cercam
Genomic Map