Assay Details
| Gene Symbol: Bfsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291061.1 | NP_001277990.1 | 1 - 2 | 600 | 139 | |
| NM_009751.2 | NP_033881.2 | 1 - 2 | 600 | 139 | ||
| GenBank mRNA | AB003147.1 | - | 1 - 2 | 370 | 139 | |
| AK053766.1 | - | 1 - 2 | 371 | 139 | ||
| AK143180.1 | - | 1 - 2 | 371 | 139 | ||
| BC057580.1 | - | 1 - 2 | 600 | 6412634 | 139 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bfsp1
Genomic Map