Assay Details
| Gene Symbol: Lamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029530.2 | NP_083806.2 | 3 - 4 | 757 | 67 | |
| GenBank mRNA | AK014127.1 | - | 1 - 2 | 199 | 67 | |
| AK018222.1 | - | 3 - 4 | 456 | 67 | ||
| AK045946.1 | - | 2 - 3 | 809 | 67 | ||
| BC004791.1 | - | 1 - 2 | 33 | 3588577 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014127.1, BC004791.1 transcripts.
Set Membership: |
|

Lamp5
Genomic Map