Assay Details
| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183113.3 | NP_898936.1 | 7 - 8 | 593 | 74 | |
| GenBank mRNA | AK016523.1 | - | 7 - 8 | 594 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016523.1, NM_183113.3 transcripts.
Set Membership: |
|

4932414N04Rik
Genomic Map