Assay Details
| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183113.3 | NP_898936.1 | 10 - 11 | 836 | 105 | |
| GenBank mRNA | AK016523.1 | - | 10 - 11 | 837 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016523.1, NM_183113.3 transcripts.
Set Membership: |
|

4932414N04Rik
Genomic Map