Assay Details
| Gene Symbol: 4932414N04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183113.3 | NP_898936.1 | 27 - 28 | 3308 | 84 | |
| GenBank mRNA | AK016523.1 | - | 27 - 28 | 3309 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

4932414N04Rik
Genomic Map