Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 23 - 24 | 2812 | 76 | |
| NM_138306.2 | NP_612179.2 | 22 - 23 | 2211 | 76 | ||
| GenBank mRNA | AK087503.1 | - | 23 - 24 | 2814 | 76 | |
| BC049228.1 | - | 22 - 23 | 2247 | 3668704 | 76 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 76 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map