Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 3 - 4 | 994 | 78 | |
| NM_138306.2 | NP_612179.2 | 2 - 3 | 393 | 78 | ||
| GenBank mRNA | AK087503.1 | - | 3 - 4 | 995 | 78 | |
| BC016217.1 | - | 2 - 3 | 1340 | 78 | ||
| BC049228.1 | - | 2 - 3 | 429 | 3668704 | 78 | |
| BY733951.1 | - | 2 - 3 | 394 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map