Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 7 - 8 | 1285 | 105 | |
| NM_138306.2 | NP_612179.2 | 6 - 7 | 684 | 105 | ||
| GenBank mRNA | AK087503.1 | - | 7 - 8 | 1286 | 105 | |
| AK170936.1 | - | 3 - 4 | 307 | 105 | ||
| AK210058.1 | - | 2 - 3 | 96 | 105 | ||
| BC016217.1 | - | 6 - 7 | 1631 | 105 | ||
| BC049228.1 | - | 6 - 7 | 720 | 3668704 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map