Assay Details
| Gene Symbol: Fubp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033389.4 | NP_001028561.2 | 14 - 15 | 1568 | 94 | |
| NM_001290548.1 | NP_001277477.1 | 14 - 15 | 1568 | 94 | ||
| GenBank mRNA | AK145377.1 | - | 14 - 15 | 94 | ||
| AK166081.1 | - | 14 - 15 | 1413 | 95 | ||
| AK166478.1 | - | 14 - 15 | 1404 | 95 | ||
| AK167671.1 | - | 14 - 15 | 1388 | 94 | ||
| BG087512.2 | - | 2 - 3 | 131 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fubp3
Genomic Map