Assay Details
| Gene Symbol: Fubp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033389.4 | NP_001028561.2 | 1 - 2 | 307 | 130 | |
| NM_001290548.1 | NP_001277477.1 | 1 - 2 | 307 | 130 | ||
| GenBank mRNA | AK137035.1 | - | 1 - 2 | 148 | 130 | |
| AK145377.1 | - | 1 - 2 | 142 | 130 | ||
| AK166081.1 | - | 1 - 2 | 150 | 130 | ||
| AK166478.1 | - | 1 - 2 | 142 | 130 | ||
| AK167671.1 | - | 1 - 2 | 127 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fubp3
Genomic Map