Assay Details
| Gene Symbol: Fam71e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172895.3 | NP_766483.3 | 9 - 10 | 2402 | 95 | |
| GenBank mRNA | AK076650.1 | - | 9 - 10 | 2461 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam71e2
Genomic Map