Assay Details
| Gene Symbol: Cdan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026891.2 | NP_081167.2 | 14 - 15 | 2200 | 87 | |
| GenBank mRNA | AK157271.1 | - | 14 - 15 | 2201 | 87 | |
| BC117555.1 | - | 12 - 13 | 1485 | 40098039 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdan1
Genomic Map