Assay Details
| Gene Symbol: Mettl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029280.4 | NP_083556.2 | 3 - 4 | 661 | 164 | |
| GenBank mRNA | AK133594.1 | - | 3 - 4 | 651 | 164 | |
| BC027547.1 | - | 3 - 4 | 594 | 1531973 | 164 | |
| BE995710.1 | - | 4 - 5 | 164 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl5
Genomic Map