Assay Details
| Gene Symbol: Nipa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY098645.1 | - | 3 - 4 | 305 | 83 | |
| BC055828.1 | - | 3 - 4 | 305 | 4192279 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nipa1
Genomic Map