Assay Details
| Gene Symbol: Pak7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172858.2 | NP_766446.2 | 5 - 6 | 1963 | 66 | |
| GenBank mRNA | AK032593.1 | - | 5 - 6 | 1936 | 66 | |
| AK079080.1 | - | 6 - 7 | 2056 | 66 | ||
| AK163530.1 | - | 5 - 6 | 1771 | 66 | ||
| AY487425.1 | - | 3 - 4 | 1483 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak7
Genomic Map