Assay Details
| Gene Symbol: Glp2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175681.3 | NP_783612.2 | 7 - 8 | 1095 | 232 | |
| GenBank mRNA | AK035702.1 | - | 490 | 232 | ||
| BC044746.2 | - | 7 - 8 | 1095 | 5363416 | 232 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glp2r
Genomic Map