Assay Details
| Gene Symbol: Rassf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175445.4 | NP_780654.1 | 5 - 6 | 646 | 61 | |
| GenBank mRNA | AK028384.1 | - | 5 - 6 | 550 | 61 | |
| AK037601.1 | - | 5 - 6 | 502 | 61 | ||
| AK172906.1 | - | 5 - 6 | 452 | 61 | ||
| BC057402.1 | - | 5 - 6 | 468 | 5698333 | 61 | |
| BQ770520.1 | - | 5 - 6 | 646 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rassf2
Genomic Map