Assay Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033431.1 | NP_001028603.1 | 6 - 7 | 2770 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp12| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033431.1 | NP_001028603.1 | 6 - 7 | 2770 | 71 | |
Set Membership: |
|