Assay Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033431.1 | NP_001028603.1 | 8 - 9 | 82 | ||
| GenBank mRNA | AK149454.1 | - | 1 - 2 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nlrp12
Genomic Map