Assay Details
| Gene Symbol: Ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025348.2 | NP_079624.1 | 3 - 4 | 187 | 76 | |
| GenBank mRNA | AK003133.1 | - | 3 - 4 | 178 | 76 | |
| AK019099.1 | - | 3 - 4 | 189 | 76 | ||
| BC061002.1 | - | 180 | 76 | |||
| BC120712.1 | - | 3 - 4 | 181 | 40129635 | 76 | |
| BC120714.1 | - | 3 - 4 | 181 | 40129637 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24946361 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184128 | AK019099.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184128 | BC061002.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184128 | NM_025348.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2184128 | AK003133.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | AF093576.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | AK161449.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | M10276.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | NM_011465.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | U87455.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339118 | XM_904631.3 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780661 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162724 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184128 | AK019099.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184128 | BC061002.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184128 | NM_025348.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2184128 | AK003133.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946373 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946377 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24946363 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162724 | - | - |
| Whole Mouse Genome G4122A | A_52_P69702 | AK019099.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69702 | BC061002.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69702 | BC120712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69702 | AK003133.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P69702 | BC120714.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115259 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162724 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24946361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24946399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946360 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24946373 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10549547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10549548 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430149_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428464_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452790_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10549546 | - | - |
| Mouse Genome 430 2.0 Array | 1452790_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430149_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452790_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1428464_at | - | - |
| Mouse Genome 430 2.0 Array | 1428464_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1430149_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66091_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66091_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700000126.mm.1 | NM_025348.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000126.mm.1 | NM_025348.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000126.mm.1 | TC0700000014.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700000127.mm.1 | NM_025348.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000127.mm.1 | NM_025348.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700000127.mm.1 | TC0700000014.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428464_PM_at | - | - |
| Mouse Expression Set 430 | 1452790_x_at | - | - |
| Mouse Expression Set 430 | 1428464_at | - | - |
| Mouse Expression Set 430 | 1452790_PM_x_at | - | - |
| Mouse Expression Set 430 | 1430149_PM_at | - | - |
| Mouse Expression Set 430 | 1430149_at | - | - |
| Murine Genome U74v2 Set | 102097_f_at | - | - |
| Murine Genome U74v2 Set | 162496_PM_r_at | - | - |
| Murine Genome U74v2 Set | 102097_PM_f_at | - | - |
| Murine Genome U74v2 Set | 96915_f_at | - | - |
| Murine Genome U74v2 Set | 162496_r_at | - | - |
| Murine Genome U74v2 Set | 96915_PM_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452790_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428464_at | - | - |
| Mouse Genome 430A 2.0 Array | 1428464_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452790_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452790_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428464_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452790_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1428464_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430149_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430149_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040438 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040438 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040438 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2772241 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2772241 | - | - |
| mouse-6 v1.1 (Illumina) | scl33146.4.49_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040438 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33146.4.49_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040438 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2792560 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2772241 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2792560 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33146.4.49_0-S | - | - |

Ndufa3
Genomic Map