Assay Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008948.2 | NP_032974.2 | 3 - 4 | 532 | 64 | |
| GenBank mRNA | AK002542.1 | - | 3 - 4 | 252 | 64 | |
| AK083043.1 | - | 494 | 64 | |||
| AK150189.1 | - | 3 - 4 | 468 | 64 | ||
| AK151370.1 | - | 3 - 4 | 252 | 64 | ||
| AK152018.1 | - | 3 - 4 | 271 | 64 | ||
| AK167002.1 | - | 3 - 4 | 480 | 64 | ||
| AK167061.1 | - | 3 - 4 | 477 | 64 | ||
| AK168263.1 | - | 2 - 3 | 201 | 64 | ||
| AK168616.1 | - | 3 - 4 | 252 | 64 | ||
| BC005783.1 | - | 3 - 4 | 421 | 3603053 | 64 | |
| CX201527.1 | - | 3 - 4 | 539 | 64 | ||
| D49686.1 | - | 3 - 4 | 476 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc3
Genomic Map