Assay Details
| Gene Symbol: Psmc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008948.2 | NP_032974.2 | 8 - 9 | 1129 | 64 | |
| GenBank mRNA | AK002542.1 | - | 8 - 9 | 850 | 64 | |
| AK083043.1 | - | 1091 | 64 | |||
| AK150189.1 | - | 8 - 9 | 1065 | 64 | ||
| AK151370.1 | - | 8 - 9 | 849 | 64 | ||
| AK152018.1 | - | 8 - 9 | 868 | 64 | ||
| AK167002.1 | - | 8 - 9 | 1077 | 64 | ||
| AK167061.1 | - | 8 - 9 | 1074 | 64 | ||
| AK168263.1 | - | 7 - 8 | 798 | 64 | ||
| AK168616.1 | - | 8 - 9 | 849 | 64 | ||
| BC005783.1 | - | 8 - 9 | 1018 | 3603053 | 64 | |
| D49686.1 | - | 8 - 9 | 1073 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmc3
Genomic Map