Assay Details
| Gene Symbol: Slc39a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290765.1 | NP_001277694.1 | 5 - 6 | 839 | 65 | |
| NM_026721.3 | NP_080997.1 | 5 - 6 | 800 | 65 | ||
| GenBank mRNA | AK035296.1 | - | 5 - 6 | 707 | 65 | |
| AK156951.1 | - | 5 - 6 | 725 | 65 | ||
| BC100463.1 | - | 5 - 6 | 807 | 30918840 | 65 | |
| BC125525.1 | - | 5 - 6 | 661 | 40130040 | 65 | |
| BC137752.1 | - | 5 - 6 | 661 | 8860771 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc39a13
Genomic Map