Assay Details
| Gene Symbol: Prpf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159714.1 | NP_001153186.1 | 1 - 2 | 70 | 71 | |
| NM_027328.4 | NP_081604.3 | 1 - 2 | 70 | 71 | ||
| GenBank mRNA | AK012987.1 | - | 1 - 2 | 64 | 71 | |
| AK033190.1 | - | 1 - 2 | 73 | 71 | ||
| AY040823.1 | - | 1 - 2 | 46 | 71 | ||
| BB839183.1 | - | 1 - 2 | 71 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK033190.1, AY040823.1, NM_001159714.1, NM_027328.4 transcripts.
Set Membership: |
|

Prpf31
Genomic Map