Assay Details
| Gene Symbol: Rars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025936.3 | NP_080212.2 | 2 - 3 | 224 | 145 | |
| GenBank mRNA | AK011383.1 | - | 2 - 3 | 202 | 145 | |
| AK076160.1 | - | 2 - 3 | 199 | 145 | ||
| AK149856.1 | - | 2 - 3 | 199 | 145 | ||
| AK152108.1 | - | 2 - 3 | 199 | 145 | ||
| AK160216.1 | - | 2 - 3 | 199 | 145 | ||
| AK166097.1 | - | 2 - 3 | 199 | 145 | ||
| AK168194.1 | - | 2 - 3 | 199 | 145 | ||
| BC020132.1 | - | 2 - 3 | 182 | 3989952 | 145 | |
| CF748175.1 | - | 2 - 3 | 225 | 146 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rars
Genomic Map